ContactUsLinkComponent
Cartridge, Nominal length: 20in, 3um Pleated Polypropylene Depth Media, SOE Fin/222t end cap with EPDM o-rings, designed for a wide range of prefiltration duties within the production of pharmaceuticals.
Catalog #: 76496-502
Supplier:  PARKER HANNIFIN TE
Cartridge, Nominal length: 20in, 3um Pleated Polypropylene Depth Media, SOE Fin/222t end cap with EPDM o-rings, designed for a wide range of prefiltration duties within the production of pharmaceuticals.
Catalog #: 76496-502
Supplier:  PARKER HANNIFIN TE
Customers Who Bought This Also Bought