Cartridge, Nominal length: 10in, 10um Pleated Polypropylene Depth Media, SOE Fin/222 end cap with EPDM o-rings, designed for a wide range of prefiltration duties within the production of pharmaceuticals.
Catalog #: 76496-494
Supplier: PARKER HANNIFIN TE
Cartridge, Nominal length: 10in, 10um Pleated Polypropylene Depth Media, SOE Fin/222 end cap with EPDM o-rings, designed for a wide range of prefiltration duties within the production of pharmaceuticals.
Catalog #: 76496-494
Supplier: PARKER HANNIFIN TE
Customers Who Bought This Also Bought
Hexane (mixture of isomers) ≥98.5% ACS
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
155,61 $ to 622,35 $
Nalgene® Cryoware Marker Set, Thermo Scientific
Supplier: Thermo Fisher Scientific
142,56 $ to 406,56 $
VWR® Laboratory Bench and Table Protectors with Leakproof Moisture Barrier
Supplier: VWR International
359,30 $
VWR® Absorbent Underpads with Waterproof Moisture Barrier
Supplier: VWR International
66,85 $ to 318,03 $
Nalgene® Clean Sheets™ Bench and Shelf Liners, Thermo Scientific
Supplier: Thermo Fisher Scientific
919,35 $ to 933,31 $
SP Bel-Art Fritware® Porous Polyethylene Sheets, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
464,49 $