Cartridge, Nominal length: 5in, 5um Pleated Polypropylene Depth Media, SOE BF/226 Bayonet end cap with EPDM o-rings, designed for a wide range of prefiltration duties within the production of pharmaceuticals.
Catalog #: 76496-364
Supplier: PARKER HANNIFIN TE
Cartridge, Nominal length: 5in, 5um Pleated Polypropylene Depth Media, SOE BF/226 Bayonet end cap with EPDM o-rings, designed for a wide range of prefiltration duties within the production of pharmaceuticals.
Catalog #: 76496-364
Supplier: PARKER HANNIFIN TE
Customers Who Bought This Also Bought
VWR® Absorbent Underpads with Waterproof Moisture Barrier
Supplier: VWR International
$240.47 to $8,653.27
VWR® Laboratory Bench and Table Protectors with Leakproof Moisture Barrier
Supplier: VWR International
$359.30
Bytac® Teflon® Resin Surface Protectors, Saint-Gobain Performance Plastics
Supplier: Saint Gobain Performance Plastics
$1,747.88
Nalgene® Clean Sheets™ Bench and Shelf Liners, Thermo Scientific
Supplier: Thermo Fisher Scientific
$919.35 to $933.31
SP Bel-Art Fritware® Porous Polyethylene Sheets, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
$464.49
Nalgene® Polyolefin Plastic Paper Sheets, Thermo Scientific
Supplier: Thermo Fisher Scientific
$534.61 to $1,523.31