Cartridge, Nominal length: 10in, 1.0um Pleated Polypropylene Depth Media, SOE BF/226 Bayonet end cap with silicone o-rings, designed for a wide range of prefiltration duties within the production of pharmaceuticals.
Catalog #: 76496-318
Supplier: PARKER HANNIFIN TE
Customers Who Bought This Also Bought