ContactUsLinkComponent
Cartridge, Nominal length: 20in, 2.0um Pleated Glass Microfibre Media, SOE BF/226 Bayonet end cap with EPDM o-rings, designed for reliable and economical removal of particulate and microorganisms from pharmaceutical fluids.
Catalog #: 76496-090
Supplier:  PARKER HANNIFIN TE
Cartridge, Nominal length: 20in, 2.0um Pleated Glass Microfibre Media, SOE BF/226 Bayonet end cap with EPDM o-rings, designed for reliable and economical removal of particulate and microorganisms from pharmaceutical fluids.
Catalog #: 76496-090
Supplier:  PARKER HANNIFIN TE
Customers Who Bought This Also Bought