ContactUsLinkComponent
Cartridge, Nominal length: 5in, 1.0um Pleated PTFE membrane, 222/Flat end cap with external Silicone o-rings, particularly suitable for sterilization and particulate removal from aggressive chemicals within a wide range of critical processing industries.
Catalog #: 76495-780
Supplier:  PARKER HANNIFIN TE
Cartridge, Nominal length: 5in, 1.0um Pleated PTFE membrane, 222/Flat end cap with external Silicone o-rings, particularly suitable for sterilization and particulate removal from aggressive chemicals within a wide range of critical processing industries.
Catalog #: 76495-780
Supplier:  PARKER HANNIFIN TE
Customers Who Bought This Also Bought