ContactUsLinkComponent
Cartridge, Nominal length: 10in, 3um Absolute Wrapped 316 Stainless Steel Media, SOE end cap with BF/226 Bayonet fitting and EPDM o-rings, ideal solution in applications where traditional polymer based filters are limited by compatibility, exposure time or high temperature and viscosity.
Catalog #: 76495-692
Supplier:  PARKER HANNIFIN TE
Cartridge, Nominal length: 10in, 3um Absolute Wrapped 316 Stainless Steel Media, SOE end cap with BF/226 Bayonet fitting and EPDM o-rings, ideal solution in applications where traditional polymer based filters are limited by compatibility, exposure time or high temperature and viscosity.
Catalog #: 76495-692
Supplier:  PARKER HANNIFIN TE
Customers Who Bought This Also Bought