ContactUsLinkComponent
Cartridge, Nominal length: 20in, .2um Pleated PTFE membrane, SOE end cap with BF/226 Bayonet fitting and external EPDM o-rings, gas sterilization filter cartridges provide unrivalled performance in process applications where continuous operation of up to 100 °C (212 °F) is a requirement.
Catalog #: 76495-606
Supplier:  PARKER HANNIFIN TE
Cartridge, Nominal length: 20in, .2um Pleated PTFE membrane, SOE end cap with BF/226 Bayonet fitting and external EPDM o-rings, gas sterilization filter cartridges provide unrivalled performance in process applications where continuous operation of up to 100 °C (212 °F) is a requirement.
Catalog #: 76495-606
Supplier:  PARKER HANNIFIN TE
Customers Who Bought This Also Bought