ContactUsLinkComponent
Cartridge, Nominal length: 10in, 0.1um Pleated PTFE membrane, Endcap: Flat Top / 222, Pharmaceutical variant, Silicone o-rings, manufactured entirely from fluoropolymers making them extremely resistant to a wide range of aggressive chemicals.
Catalog #: 76495-450
Supplier:  PARKER HANNIFIN TE
Cartridge, Nominal length: 10in, 0.1um Pleated PTFE membrane, Endcap: Flat Top / 222, Pharmaceutical variant, Silicone o-rings, manufactured entirely from fluoropolymers making them extremely resistant to a wide range of aggressive chemicals.
Catalog #: 76495-450
Supplier:  PARKER HANNIFIN TE
Customers Who Bought This Also Bought