ContactUsLinkComponent
Cartridge, Nominal length: 20in, 0.1um Pleated PTFE membrane, Endcap: BF / 226 Bayonet, Pharmaceutical variant, Silicone o-rings, manufactured entirely from fluoropolymers making them extremely resistant to a wide range of aggressive chemicals.
Catalog #: 76495-362
Supplier:  PARKER HANNIFIN TE
Cartridge, Nominal length: 20in, 0.1um Pleated PTFE membrane, Endcap: BF / 226 Bayonet, Pharmaceutical variant, Silicone o-rings, manufactured entirely from fluoropolymers making them extremely resistant to a wide range of aggressive chemicals.
Catalog #: 76495-362
Supplier:  PARKER HANNIFIN TE
Customers Who Bought This Also Bought