ContactUsLinkComponent
Cartridge, Nominal length: 20in, 0.1um Pleated PTFE membrane, SOE end cap with BF / 226 Bayonet fitting and Pharmaceutical variant, FEP encapsulated Viton o-rings, manufactured entirely from fluoropolymers making them extremely resistant to a wide range of aggressive chemicals.
Catalog #: 76495-338
Supplier:  PARKER HANNIFIN TE
Cartridge, Nominal length: 20in, 0.1um Pleated PTFE membrane, SOE end cap with BF / 226 Bayonet fitting and Pharmaceutical variant, FEP encapsulated Viton o-rings, manufactured entirely from fluoropolymers making them extremely resistant to a wide range of aggressive chemicals.
Catalog #: 76495-338
Supplier:  PARKER HANNIFIN TE
Customers Who Bought This Also Bought