ContactUsLinkComponent
Cartridge, Insert Style: No insert, Standard, End Fitting: 222/Fin, Nominal Length: 30in, Filter Rating: 0.65/0.2um, Gasket / O-Rings: EPDM, Membrane Type: 0.45/0.2um, Depth media: Pleated PP, provides excellent retention efficiencies/onstream life, optimized for use in liquid applications
Catalog #: 76495-050
Supplier:  PARKER HANNIFIN TE
Cartridge, Insert Style: No insert, Standard, End Fitting: 222/Fin, Nominal Length: 30in, Filter Rating: 0.65/0.2um, Gasket / O-Rings: EPDM, Membrane Type: 0.45/0.2um, Depth media: Pleated PP, provides excellent retention efficiencies/onstream life, optimized for use in liquid applications
Catalog #: 76495-050
Supplier:  PARKER HANNIFIN TE
Customers Who Bought This Also Bought