ContactUsLinkComponent
Cartridge, Insert Style: No insert, Standard, End Fitting: DOE, Nominal Length: 20in, Filter Rating: 0.65/0.2um, Gasket/Orings: Silicone, Membrane Type: 0.65/0.2um, Depth media: Pleated Polypropylene, provides excellent retention efficiencies/onstream life, optimized for use in liquid applications
Catalog #: 76495-006
Supplier:  PARKER HANNIFIN TE
Cartridge, Insert Style: No insert, Standard, End Fitting: DOE, Nominal Length: 20in, Filter Rating: 0.65/0.2um, Gasket/Orings: Silicone, Membrane Type: 0.65/0.2um, Depth media: Pleated Polypropylene, provides excellent retention efficiencies/onstream life, optimized for use in liquid applications
Catalog #: 76495-006
Supplier:  PARKER HANNIFIN TE
Customers Who Bought This Also Bought