ContactUsLinkComponent
Cartridge, Insert Style: No insert, Standard, End Fitting: 222/Fin, Nominal Length: 10in, Filter Rating: 0.45/0.2um, Gasket/O-Rings: EPDMMembrane type: 0.45/0.2um, Depth media: Pleated Polypropylene, provides excellent retention efficiencies/onstream life, optimized for use in liquid applications
Catalog #: 76494-986
Supplier:  PARKER HANNIFIN TE
Cartridge, Insert Style: No insert, Standard, End Fitting: 222/Fin, Nominal Length: 10in, Filter Rating: 0.45/0.2um, Gasket/O-Rings: EPDMMembrane type: 0.45/0.2um, Depth media: Pleated Polypropylene, provides excellent retention efficiencies/onstream life, optimized for use in liquid applications
Catalog #: 76494-986
Supplier:  PARKER HANNIFIN TE
Customers Who Bought This Also Bought