ContactUsLinkComponent
Cartridge, Insert Style: No insert, Standard, End Fitting: 226/Fin, Nominal Length: 30in, Filter Rating: 0.8um, O-ring: Silicone, Depth media: Pleated Polypropylene, provides excellent retention efficiencies and onstream life, optimized for use in liquid applications
Catalog #: 76494-868
Supplier:  PARKER HANNIFIN TE
Cartridge, Insert Style: No insert, Standard, End Fitting: 226/Fin, Nominal Length: 30in, Filter Rating: 0.8um, O-ring: Silicone, Depth media: Pleated Polypropylene, provides excellent retention efficiencies and onstream life, optimized for use in liquid applications
Catalog #: 76494-868
Supplier:  PARKER HANNIFIN TE
Customers Who Bought This Also Bought