ContactUsLinkComponent
Cartridge, Insert Style: No insert, Standard, End Fitting: 222/Fin, Nominal Length: 5in, Filter Rating: 3um, Gasket/O-Rings: Silicone, Thickness: 0.200in (5 mm), Depth media: Pleated Polypropylene, provides excellent retention efficiencies/onstream life, optimized for use in liquid applications
Catalog #: 76494-632
Supplier:  PARKER HANNIFIN TE
Cartridge, Insert Style: No insert, Standard, End Fitting: 222/Fin, Nominal Length: 5in, Filter Rating: 3um, Gasket/O-Rings: Silicone, Thickness: 0.200in (5 mm), Depth media: Pleated Polypropylene, provides excellent retention efficiencies/onstream life, optimized for use in liquid applications
Catalog #: 76494-632
Supplier:  PARKER HANNIFIN TE
Customers Who Bought This Also Bought