ContactUsLinkComponent
Cartridge, Insert Style: No insert, Standard, End Fitting: 222/Flat, Nominal Length: 20in, Filter Rating: 1um, Gasket O-ring: EPDM, Thickness: 0.200in (5 mm), Depth media: Pleated Polypropylene, provides excellent retention efficiencies and onstream life, optimized for use in liquid applications
Catalog #: 76494-450
Supplier:  PARKER HANNIFIN TE
Cartridge, Insert Style: No insert, Standard, End Fitting: 222/Flat, Nominal Length: 20in, Filter Rating: 1um, Gasket O-ring: EPDM, Thickness: 0.200in (5 mm), Depth media: Pleated Polypropylene, provides excellent retention efficiencies and onstream life, optimized for use in liquid applications
Catalog #: 76494-450
Supplier:  PARKER HANNIFIN TE
Customers Who Bought This Also Bought