ContactUsLinkComponent
Cartridge, Insert Style: No insert, Standard, End Fitting: DOE, Nominal Length: 10in, Filter Rating: 0.5um, Gasket/O-Rings: Silicone, Thickness: 0.200in (5 mm), Depth media: Pleated Polypropylene, provides excellent retention efficiencies and onstream life, optimized for use in liquid applications
Catalog #: 76494-318
Supplier:  PARKER HANNIFIN TE
Cartridge, Insert Style: No insert, Standard, End Fitting: DOE, Nominal Length: 10in, Filter Rating: 0.5um, Gasket/O-Rings: Silicone, Thickness: 0.200in (5 mm), Depth media: Pleated Polypropylene, provides excellent retention efficiencies and onstream life, optimized for use in liquid applications
Catalog #: 76494-318
Supplier:  PARKER HANNIFIN TE
Customers Who Bought This Also Bought