Meiv Safety Glasses Clear Lens In Clam shell, Ultra lightweight metal alloy frame features eight base spherical polycarbonate lenses with an abrasion resistant coating that provides 99.9% UV protection. Adjustable soft PVC nose pads and rubberized temple tips
Catalog #: CA76492-938
Supplier: Dentec Safety Specialists
Customers Who Bought This Also Bought