Miranda Safety Glasses Clear Af Lens Cl, Features soft PVC nose pads, spatula style temples with rubberized tips, and impact resistant poly- carbonate lens that provides 99.9% UV protection. Includes a scratch resistant coating to prolong the life of the lens
Catalog #: CA76492-900
Supplier: Dentec Safety Specialists
Customers Who Bought This Also Bought