AAA Guard Column AssemblyGuard Column Housing And One (1) Guard Column Cartridge For L-8800/L-8900/La8080. * Includes All Necessary Tools, Tubing And Fittings.
Catalog #: 76493-020
Supplier: HITACHI HIGH-TECH AMERICA, INC.
AAA Guard Column AssemblyGuard Column Housing And One (1) Guard Column Cartridge For L-8800/L-8900/La8080. * Includes All Necessary Tools, Tubing And Fittings.
Catalog #: 76493-020
Supplier: HITACHI HIGH-TECH AMERICA, INC.
Customers Who Bought This Also Bought
Glycerine ≥99.7%, Laboratory Reagent, VWR Chemicals BDH®
Supplier: VWR International
$881.80 to $2,807.56
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$635.20 to $2,541.50
VRB (Violet Red Bile) Agar, Fluorocult®, Dehydrated Culture Media, MilliporeSigma
Supplier: MilliporeSigma
$1,692.83
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$684.63 to $2,738.51
Tetrahydrofuran, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Supplier: VWR International
$480.10 to $2,245.11
Acetone ≥99.5% (by GC, corrected for water content), OmniSolv®, Supelco®
Supplier: MilliporeSigma
$768.73 to $3,075.66
Formaldehyde (Paraformaldehyde) 16% in aqueous solution, EM Grade
Supplier: Electron Microscopy Science
$104.49