ContactUsLinkComponent
Kit, First Aid On Section 9 Metal, For Ontario workplaces with 6 to 15 workers at any given time.
First Aid Central Ontario First Aid Kits, Acme United
Catalog #: CA76491-924
Kit, First Aid On Section 9 Metal, For Ontario workplaces with 6 to 15 workers at any given time.
First Aid Central Ontario First Aid Kits, Acme United
Catalog #: CA76491-924
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Ontario section 9 first aid kit, in metal case
  • Cat. no.:
    CA76491-924
Specifications
About this item
Economical and high-quality Ontario Regulation First Aid Kits meet Ontario OHS requirements.
  • Designed specifically for workplaces and industries following Ontario first aid requirements
  • Kits meet Ontario WSIB standards for workplace first aid kits
  • Includes a variety of first aid supplies for treating common workplace injuries
  • Available in nylon bag, wall mountable plastic, and metal case options
Employers in Ontario that are covered by the Workplace Safety and Insurance Act are required to equip their workplaces with first aid kits that meet the Occupational Health and Safety Act’s Regulation 1101 first aid kit requirements. The type of Ontario first aid kit required is determined by the number of employees on shift at any given moment, as outlined in the following guidelines.
First aid requirements vary depending on the situation such as: Ontario Section 8 First Aid Kit – for workplaces with 1-5 workers per shift; Ontario Section 9 First Aid Kit – for workplaces with 6-15 workers per shift; Ontario Section 10 First Aid Kit – for workplaces with 16-199 workers per shift; Ontario Section 16 First Aid Kit – for a workplace vehicle used to transport workers or goods.
Customers Who Bought This Also Bought