VWR® Environmentally Friendly Thermometers, Easy Read
Catalog #: 76491-596
Supplier: VWR International
Specifications
- Accuracy:±1.0, ±1.5 (>100 °C)
- Length:200 mm
- Cat. no.:76491-596
- Divisions:1.0 °C
- Range:−10...+150 °C
- Immersion:50 mm
Specifications
About this item
These thermometers are easy to read quickly and accurately due to the high contrast black numbering against a yellow background. Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.
- Enviro safe liquid
- Eliminates reading errors
- Non-toxic, non-hazardous, biodegradable liquid
Includes multi-language (English, French, German, Italian, Portuguese and Spanish) statement of accuracy and traceability, non roll fitting and individual serial number.
Certifications: Traceable NIST. Envirokleen certified by ChemTel (an independent laboratory).
Customers Who Bought This Also Bought