Reagent Reservoirs, Globe Scientific
Catalog #: 76491-560
Supplier: Globe Scientific
Specifications
- Capacity:10 ml
- Length:102 mm (4.01")
- Sterility:Sterile
- Width:51.5 mm (2.02")
- Color:White
- Height:21.3 mm (0.84")
- Packaging:Individually wrapped
Specifications
About this item
These reagent reservoirs are suitable for virtually all of your lab’s sample preparation and aliquoting needs.
- Pour spouts at every corner
- Rigid sidewall design
- Wide, stable base
- Molded graduations
- Compatible with multi-channel pipettors
- Dnase, Rnase, purogen, animal materials and latex free
- Produced from virgin medical grade polystyrene
Design enhancements include pour spouts at each corner, a rigid sidewall design and graduations for easy volume reference. Sterile variants are irradiated to guarantee a sterility assurance level (SAL) of 10-⁶. Available volumes of 10 ml, 25 ml and 50 ml ensure there is a volume and packaging option to fit almost every application. Individual, bulk and five-pack options are available.
Customers Who Bought This Also Bought