ContactUsLinkComponent
Tube, Test, Reusable, Beaded Rim, 4mL, 13mm x 55mm (OD x H) ISO/DIN 4142, Thick walled, Specifications: Material: 3.3 Borosilicate, Color: Clear, Capacity: 4mL, Approx O.D. X Length: 13mm x 55mm, Class/ Quality Grade: Type I,
Catalog #: 76491-522
Supplier:  Foxx Life Sciences
Tube, Test, Reusable, Beaded Rim, 4mL, 13mm x 55mm (OD x H) ISO/DIN 4142, Thick walled, Specifications: Material: 3.3 Borosilicate, Color: Clear, Capacity: 4mL, Approx O.D. X Length: 13mm x 55mm, Class/ Quality Grade: Type I,
Tube, Test, Reusable, Beaded Rim, 4mL, 13mm x 55mm (OD x H) ISO/DIN 4142, Thick walled, Specifications: Material: 3.3 Borosilicate, Color: Clear, Capacity: 4mL, Approx O.D. X Length: 13mm x 55mm, Class/ Quality Grade: Type I,
Catalog #: 76491-522
Supplier:  Foxx Life Sciences
Customers Who Bought This Also Bought