ContactUsLinkComponent
Test Tube with Rim, ISO/DIN 4142, thick walled, 100mL, Manufacturer:, Quantity: Case of 50, SKU/Product Code: 9800U10, Material: 3.3 Borosilicate, Color: Clear, Capacity: 100mL, Size/Dimensions: Approx O.D. X Length (EA): 32mm x 200mm (1.25 x 7.87in), Weight: 10.15 lbs, Grade: Type I
Catalog #: 76491-518
Supplier:  Foxx Life Sciences
Test Tube with Rim, ISO/DIN 4142, thick walled, 100mL, Manufacturer:, Quantity: Case of 50, SKU/Product Code: 9800U10, Material: 3.3 Borosilicate, Color: Clear, Capacity: 100mL, Size/Dimensions: Approx O.D. X Length (EA): 32mm x 200mm (1.25 x 7.87in), Weight: 10.15 lbs, Grade: Type I
Test Tube with Rim, ISO/DIN 4142, thick walled, 100mL, Manufacturer:, Quantity: Case of 50, SKU/Product Code: 9800U10, Material: 3.3 Borosilicate, Color: Clear, Capacity: 100mL, Size/Dimensions: Approx O.D. X Length (EA): 32mm x 200mm (1.25 x 7.87in), Weight: 10.15 lbs, Grade: Type I
Catalog #: 76491-518
Supplier:  Foxx Life Sciences
Customers Who Bought This Also Bought