ContactUsLinkComponent
Tube, Test, Reusable, Beaded Rim, 70mL, 25mm x 200mm (OD x H) ISO/DIN 4142, Thick walled, Specifications: Material: 3.3 Borosilicate, Color: Clear, Capacity: 70mL, Approx O.D. X Length: 25mm x 200mm, Class/ Quality Grade: Type I,
Catalog #: 76491-516
Supplier:  Foxx Life Sciences
Tube, Test, Reusable, Beaded Rim, 70mL, 25mm x 200mm (OD x H) ISO/DIN 4142, Thick walled, Specifications: Material: 3.3 Borosilicate, Color: Clear, Capacity: 70mL, Approx O.D. X Length: 25mm x 200mm, Class/ Quality Grade: Type I,
Tube, Test, Reusable, Beaded Rim, 70mL, 25mm x 200mm (OD x H) ISO/DIN 4142, Thick walled, Specifications: Material: 3.3 Borosilicate, Color: Clear, Capacity: 70mL, Approx O.D. X Length: 25mm x 200mm, Class/ Quality Grade: Type I,
Catalog #: 76491-516
Supplier:  Foxx Life Sciences
Customers Who Bought This Also Bought