Test Tube with Rim 15*125 icate ISO/DIN 4142, thick walled, CS/800, 13 mL, Specifications: Material: Glass, Volumen: 13 mL, Overall Dimension: 15mm x 125mm, icate Glass 3.3, Graduation Range: Type I, Documentation, Datasheets Coming Soon!
Catalog #: 76491-506
Supplier: Foxx Life Sciences
Customers Who Bought This Also Bought