ContactUsLinkComponent
Test Tube with Rim 12*100 icate ISO/DIN 4142, thick walled, CS/800, 7 mL, Specifications: Material: Glass, Volumen: 7 mL, Overall Dimension: 12mm x 100mm, icate Glass 3.3, Graduation Range: Type I, Documentation, Datasheets Coming Soon!
Catalog #: 76491-504
Supplier:  Foxx Life Sciences
Test Tube with Rim 12*100 icate ISO/DIN 4142, thick walled, CS/800, 7 mL, Specifications: Material: Glass, Volumen: 7 mL, Overall Dimension: 12mm x 100mm, icate Glass 3.3, Graduation Range: Type I, Documentation, Datasheets Coming Soon!
Test Tube with Rim 12*100 icate ISO/DIN 4142, thick walled, CS/800, 7 mL, Specifications: Material: Glass, Volumen: 7 mL, Overall Dimension: 12mm x 100mm, icate Glass 3.3, Graduation Range: Type I, Documentation, Datasheets Coming Soon!
Catalog #: 76491-504
Supplier:  Foxx Life Sciences
Customers Who Bought This Also Bought