ContactUsLinkComponent
Penny Head Stopper, Amber, Made With Amber Solid Penny Head Glass Stopper, Interchangeable Ground Joint 24/29, Specifications: Material: 3.3 Borosilicate, Color: Amber, Class/Quality Grade: Class A/Type I, Dimensions: 155mm(6.10in) X 50mm(1.97in) X 155mm(6.10in), Joint size: 24/29,
Catalog #: 76491-494
Supplier:  Foxx Life Sciences
Penny Head Stopper, Amber, Made With Amber Solid Penny Head Glass Stopper, Interchangeable Ground Joint 24/29, Specifications: Material: 3.3 Borosilicate, Color: Amber, Class/Quality Grade: Class A/Type I, Dimensions: 155mm(6.10in) X 50mm(1.97in) X 155mm(6.10in), Joint size: 24/29,
Penny Head Stopper, Amber, Made With Amber Solid Penny Head Glass Stopper, Interchangeable Ground Joint 24/29, Specifications: Material: 3.3 Borosilicate, Color: Amber, Class/Quality Grade: Class A/Type I, Dimensions: 155mm(6.10in) X 50mm(1.97in) X 155mm(6.10in), Joint size: 24/29,
Catalog #: 76491-494
Supplier:  Foxx Life Sciences
Customers Who Bought This Also Bought