Penny Head Stopper, Amber, Made With Amber Solid Penny Head Glass Stopper, Interchangeable Ground Joint 14/23, Specifications: Material: 3.3 Borosilicate, Color: Amber, Class/Quality Grade: Class A/Type I, Dimensions: 136mm(5.35in) X 72mm(2.83in) X 120mm(4.72in), Joint size: 14/23,
Catalog #: 76491-492
Supplier: Foxx Life Sciences
Customers Who Bought This Also Bought