ContactUsLinkComponent
Volumetric Transfer Pipette Class A Accuracy ASTM E-969-02 One Mark, Bulb Pipette, with Batch Certificate icate 3.3 USP Type I, 100 mL, Specifications: Material: 3.3 Borosilicate Glass, Color: Clear, Capacity: 100 mL, Class/ Quality Grade: Type I, Class A, Tolerance: 0.08 mL,
Catalog #: 76491-436
Supplier:  Foxx Life Sciences
Volumetric Transfer Pipette Class A Accuracy ASTM E-969-02 One Mark, Bulb Pipette, with Batch Certificate icate 3.3 USP Type I, 100 mL, Specifications: Material: 3.3 Borosilicate Glass, Color: Clear, Capacity: 100 mL, Class/ Quality Grade: Type I, Class A, Tolerance: 0.08 mL,
Volumetric Transfer Pipette Class A Accuracy ASTM E-969-02 One Mark, Bulb Pipette, with Batch Certificate icate 3.3 USP Type I, 100 mL, Specifications: Material: 3.3 Borosilicate Glass, Color: Clear, Capacity: 100 mL, Class/ Quality Grade: Type I, Class A, Tolerance: 0.08 mL,
Catalog #: 76491-436
Supplier:  Foxx Life Sciences
Customers Who Bought This Also Bought