ContactUsLinkComponent
Volumetric Transfer Pipette Class A Accuracy ASTM E-969-02 with Batch Certificate 5mL icate 3.3, Material: 3.3 Borosilicate Glass, Color: Clear, Capacity: 5mL, Overall Dimension: 18.70in L x 6.89in W x 2.95in H, Tolerance:.01mL, Class/Quality Grade: Type 1, Class A,
Catalog #: 76491-420
Supplier:  Foxx Life Sciences
Volumetric Transfer Pipette Class A Accuracy ASTM E-969-02 with Batch Certificate 5mL icate 3.3, Material: 3.3 Borosilicate Glass, Color: Clear, Capacity: 5mL, Overall Dimension: 18.70in L x 6.89in W x 2.95in H, Tolerance:.01mL, Class/Quality Grade: Type 1, Class A,
Volumetric Transfer Pipette Class A Accuracy ASTM E-969-02 with Batch Certificate 5mL icate 3.3, Material: 3.3 Borosilicate Glass, Color: Clear, Capacity: 5mL, Overall Dimension: 18.70in L x 6.89in W x 2.95in H, Tolerance:.01mL, Class/Quality Grade: Type 1, Class A,
Catalog #: 76491-420
Supplier:  Foxx Life Sciences
Customers Who Bought This Also Bought