ContactUsLinkComponent
Volumetric Transfer Pipette Class A Accuracy ASTM E-969-02 with Batch Certificate 2mL icate 3.3, Material: 3.3 Borosilicate Glass, Color: Clear, Capacity: 2mL, Overall Dimension: 14.96in L x 5.12in W x 2.17in H, Tolerance:.006mL, Class/Quality Grade: Type 1, Class A,
Catalog #: 76491-414
Supplier:  Foxx Life Sciences
Volumetric Transfer Pipette Class A Accuracy ASTM E-969-02 with Batch Certificate 2mL icate 3.3, Material: 3.3 Borosilicate Glass, Color: Clear, Capacity: 2mL, Overall Dimension: 14.96in L x 5.12in W x 2.17in H, Tolerance:.006mL, Class/Quality Grade: Type 1, Class A,
Volumetric Transfer Pipette Class A Accuracy ASTM E-969-02 with Batch Certificate 2mL icate 3.3, Material: 3.3 Borosilicate Glass, Color: Clear, Capacity: 2mL, Overall Dimension: 14.96in L x 5.12in W x 2.17in H, Tolerance:.006mL, Class/Quality Grade: Type 1, Class A,
Catalog #: 76491-414
Supplier:  Foxx Life Sciences
Customers Who Bought This Also Bought