Flask, Boiling, Round Bottom, Ground Glass Neck, 50mL, 24/29 Interchangeable Joint, Specifications: Material: 3.3 Borosilicate, Color: Clear, Capacity: 50mL, Approx Height: 24/29, Interchangeable Joint: 51mm x 105mm, Class/ Quality Grade: Type I,
Catalog #: 76491-108
Supplier: Foxx Life Sciences
Customers Who Bought This Also Bought