ContactUsLinkComponent
Burette, Boroflo fitted with Boroflo General Purpose Screw Thread PTFE Keys, Accuracy as per Class B, 50 mL, 3.3 Borosilicate Glass, Color: Clear, Capacity: 50mL, Graduation Range: 0- 50mL, Graduation Interval: 0.10mL, Graduation Color: Ceramic White, Tolerance: 0.10mL, Grade: Type I, Class B,
Catalog #: 76491-070
Supplier:  Foxx Life Sciences
Burette, Boroflo fitted with Boroflo General Purpose Screw Thread PTFE Keys, Accuracy as per Class B, 50 mL, 3.3 Borosilicate Glass, Color: Clear, Capacity: 50mL, Graduation Range: 0- 50mL, Graduation Interval: 0.10mL, Graduation Color: Ceramic White, Tolerance: 0.10mL, Grade: Type I, Class B,
Burette, Boroflo fitted with Boroflo General Purpose Screw Thread PTFE Keys, Accuracy as per Class B, 50 mL, 3.3 Borosilicate Glass, Color: Clear, Capacity: 50mL, Graduation Range: 0- 50mL, Graduation Interval: 0.10mL, Graduation Color: Ceramic White, Tolerance: 0.10mL, Grade: Type I, Class B,
Catalog #: 76491-070
Supplier:  Foxx Life Sciences
Customers Who Bought This Also Bought