ContactUsLinkComponent
Burette with Batch Certificate as per ASTM E 287, Specifications: Material: 3.3 Borosilicate Glass , Capacity: 50mL, Color: Clear, Overall Dimension: 32.83in L x 6.50in W x 9.61in H, Graduation Color: Ceramic White, Class/Quality Grade: Type 1, Class A , Documentation, Burette Data Sheet
Catalog #: 76491-058
Supplier:  Foxx Life Sciences
Burette with Batch Certificate as per ASTM E 287, Specifications: Material: 3.3 Borosilicate Glass , Capacity: 50mL, Color: Clear, Overall Dimension: 32.83in L x 6.50in W x 9.61in H, Graduation Color: Ceramic White, Class/Quality Grade: Type 1, Class A , Documentation, Burette Data Sheet
Burette with Batch Certificate as per ASTM E 287, Specifications: Material: 3.3 Borosilicate Glass , Capacity: 50mL, Color: Clear, Overall Dimension: 32.83in L x 6.50in W x 9.61in H, Graduation Color: Ceramic White, Class/Quality Grade: Type 1, Class A , Documentation, Burette Data Sheet
Catalog #: 76491-058
Supplier:  Foxx Life Sciences
Customers Who Bought This Also Bought