ContactUsLinkComponent
Burette with Batch Certificate as per ASTM E 287, Specifications: Material: 3.3 Borosilicate Glass , Capacity: 25mL, Color: Clear, Overall Dimension: 24.02in L x 16.14in W x 3.35in H, Graduation Color: Ceramic White, Class/Quality Grade: Type 1, Class A , Documentation, Burette Data Sheet
Catalog #: 76491-054
Supplier:  Foxx Life Sciences
Burette with Batch Certificate as per ASTM E 287, Specifications: Material: 3.3 Borosilicate Glass , Capacity: 25mL, Color: Clear, Overall Dimension: 24.02in L x 16.14in W x 3.35in H, Graduation Color: Ceramic White, Class/Quality Grade: Type 1, Class A , Documentation, Burette Data Sheet
Burette with Batch Certificate as per ASTM E 287, Specifications: Material: 3.3 Borosilicate Glass , Capacity: 25mL, Color: Clear, Overall Dimension: 24.02in L x 16.14in W x 3.35in H, Graduation Color: Ceramic White, Class/Quality Grade: Type 1, Class A , Documentation, Burette Data Sheet
Catalog #: 76491-054
Supplier:  Foxx Life Sciences
Customers Who Bought This Also Bought