ContactUsLinkComponent
Media Amber Culture Tube, Round Bottom, PP Screw Cap with PTFE Liner, 150 mL, Material: 3.3 Borosilicate Glass, Color: Amber, Capacity: 150mL, Approx O.D. X Length: 38mm x 200mm, Class/ Quality Grade: Type I, Class A,
Catalog #: 76491-010
Supplier:  Foxx Life Sciences
Media Amber Culture Tube, Round Bottom, PP Screw Cap with PTFE Liner, 150 mL, Material: 3.3 Borosilicate Glass, Color: Amber, Capacity: 150mL, Approx O.D. X Length: 38mm x 200mm, Class/ Quality Grade: Type I, Class A,
Catalog #: 76491-010
Supplier:  Foxx Life Sciences
Customers Who Bought This Also Bought