ContactUsLinkComponent
Culture Tube Media Round Bottom PP cap with Liner 50 mL icate CS/50, Specifications: Material: 3.3 Borosilicate Glass, Tube size: 50 mL, Color: Clear, Cap Material: Polypropylene, Overall Dimension: 9900012, Class/Quality Grade:  Type I, Class A, Documentation, Datasheets Coming Soon!
Catalog #: 76490-996
Supplier:  Foxx Life Sciences
Culture Tube Media Round Bottom PP cap with Liner 50 mL icate CS/50, Specifications: Material: 3.3 Borosilicate Glass, Tube size: 50 mL, Color: Clear, Cap Material: Polypropylene, Overall Dimension: 9900012, Class/Quality Grade:  Type I, Class A, Documentation, Datasheets Coming Soon!
Culture Tube Media Round Bottom PP cap with Liner 50 mL icate CS/50, Specifications: Material: 3.3 Borosilicate Glass, Tube size: 50 mL, Color: Clear, Cap Material: Polypropylene, Overall Dimension: 9900012, Class/Quality Grade:  Type I, Class A, Documentation, Datasheets Coming Soon!
Catalog #: 76490-996
Supplier:  Foxx Life Sciences
Customers Who Bought This Also Bought