ContactUsLinkComponent
Amber Solid Penny Head Glass Stopper, Interchangeable Ground Joint 55/54, Specifications: Material: 3.3 Borosilicate, Color: Amber, Neck Type: Stopper, Joint Size: 55/54, Class/ Quality Grade: Type I, Class A,
Catalog #: 76490-960
Supplier:  Foxx Life Sciences
Amber Solid Penny Head Glass Stopper, Interchangeable Ground Joint 55/54, Specifications: Material: 3.3 Borosilicate, Color: Amber, Neck Type: Stopper, Joint Size: 55/54, Class/ Quality Grade: Type I, Class A,
Amber Solid Penny Head Glass Stopper, Interchangeable Ground Joint 55/54, Specifications: Material: 3.3 Borosilicate, Color: Amber, Neck Type: Stopper, Joint Size: 55/54, Class/ Quality Grade: Type I, Class A,
Catalog #: 76490-960
Supplier:  Foxx Life Sciences
Customers Who Bought This Also Bought