ContactUsLinkComponent
Glass Separating Funnel Pear Shape, Boroflo Stopcock with PTFE key, Interchangeable Glass Stopper, USP Type I icate 3.3, ISO 4800, 60 mL, Material: 3.3 Borosilicate Glass, Color: Clear, Capacity: 60mL, Stopper Size: 14/23, Class/ Quality Grade: Type I,
Catalog #: 76490-850
Supplier:  Foxx Life Sciences
Glass Separating Funnel Pear Shape, Boroflo Stopcock with PTFE key, Interchangeable Glass Stopper, USP Type I icate 3.3, ISO 4800, 60 mL, Material: 3.3 Borosilicate Glass, Color: Clear, Capacity: 60mL, Stopper Size: 14/23, Class/ Quality Grade: Type I,
Glass Separating Funnel Pear Shape, Boroflo Stopcock with PTFE key, Interchangeable Glass Stopper, USP Type I icate 3.3, ISO 4800, 60 mL, Material: 3.3 Borosilicate Glass, Color: Clear, Capacity: 60mL, Stopper Size: 14/23, Class/ Quality Grade: Type I,
Catalog #: 76490-850
Supplier:  Foxx Life Sciences
Customers Who Bought This Also Bought