ContactUsLinkComponent
Vacuum icate Glass Filtration Assembly 47MM, 1L flask 300mL funnel with an aluminum clamp, Glass, Flask Volume: 1L, Funnel Cup Volume: 300mL, Membrane Holder Diameter: 47mm, Clamp material: Aluminum, Overall Dimension: 7.68in L x 7.68in W x 8.07in H,
Catalog #: 76490-818
Supplier:  Foxx Life Sciences
Vacuum icate Glass Filtration Assembly 47MM, 1L flask 300mL funnel with an aluminum clamp, Glass, Flask Volume: 1L, Funnel Cup Volume: 300mL, Membrane Holder Diameter: 47mm, Clamp material: Aluminum, Overall Dimension: 7.68in L x 7.68in W x 8.07in H,
Vacuum icate Glass Filtration Assembly 47MM, 1L flask 300mL funnel with an aluminum clamp, Glass, Flask Volume: 1L, Funnel Cup Volume: 300mL, Membrane Holder Diameter: 47mm, Clamp material: Aluminum, Overall Dimension: 7.68in L x 7.68in W x 8.07in H,
Catalog #: 76490-818
Supplier:  Foxx Life Sciences
Customers Who Bought This Also Bought