Flask, Filtering, Beaded Rim, 10LSpecifications: Material: 3.3 Borosilicate Glass, Color: Clear, Capacity: 10L, Approx O.D. X Height: 230mm x 488mm, Class/ Quality Grade: Type I, neck finish is developed for strength and to ensure a proper stopper fitting
Catalog #: 76490-812
Supplier: Foxx Life Sciences
Customers Who Bought This Also Bought