Filtration Flask Bolt Neck with Tubulation, 3.3 Borosilicate, 250mL, Material: 3.3 Borosilicate Glass, Color: Clear, Capacity: 250mL, Approx O.D. X Height: 85mm x 155mm, Class/ Quality Grade: Type I,
Catalog #: 76490-802
Supplier: Foxx Life Sciences
Customers Who Bought This Also Bought