Filtration Flask Bolt Neck with Tubulation, 3.3 Borosilicate, 100mL, Material: 3.3 Borosilicate Glass, Color: Clear, Capacity: 100mL, Approx O.D. X Height: 64mm x 108mm, Class/ Quality Grade: Type I,
Catalog #: 76490-800
Supplier: Foxx Life Sciences
Customers Who Bought This Also Bought