ContactUsLinkComponent
Kit, First Aid Csa Type 3 Sm Refill, For high risk workplaces following CSA standards and having 2-25 workers per shift.
First Aid Central CSA First Aid Kits, Acme United
Catalog #: CA76491-794
Kit, First Aid Csa Type 3 Sm Refill, For high risk workplaces following CSA standards and having 2-25 workers per shift.
First Aid Central CSA First Aid Kits, Acme United
Catalog #: CA76491-794
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    CSA type 3 intermediate first aid kit, small, refill
  • Cat. no.:
    CA76491-794
Specifications
About this item
Economical and high-quality CSA First Aid Kits meet CSA Z1220-17 standards.
  • Designed specifically for workplaces and industries following CSA first aid standards
  • Kits meet CSA Z1220-17 standards for workplace first aid kits
  • Includes a variety of first aid supplies for treating common workplace injuries
  • Available in nylon bag, wall mountable plastic, and metal case options
Canadian Standards Association CSA Z1220-17 first aid kits for the workplace are required for the provinces of MB, NB, NL, NWT, NU, QC, and SK and are accepted in Ontario as per WSIB requirements. The type of CSA first aid kit required for a workplace is determined by the number of employees per shift as well as the level of risk of injury in the workplace. First Aid Central’s complete line of CSA first aid kits help keep employees safe and ensure that CSA first aid requirements for the workplace are met.
Lone/isolated worker: 1 worker or worker in isolation requires a CSA Type 1 Personal First Aid Kit.
For low risk workplace environment with: 2-25 workers per shift requires a CSA Type 2 Basic First Aid Kit - Small (2-25 Workers); 26-50 workers per shift requires a CSA Type 2 Basic First Aid Kit - Medium (26-50 Workers); 51-100 workers per shift requires a CSA Type 2 Basic First Aid Kit - Large (51-100 Workers).
For high risk workplace environment with: 2-25 workers per shift requires a CSA Type 3 Intermediate First Aid Kit - Small (2-25 Workers); 26-50 workers per shift requires a CSA Type 3 Intermediate First Aid Kit - Medium (26-50 Workers); 51-100 workers per shift requires a CSA Type 3 Intermediate First Aid Kit - Large (51-100 Workers).
Customers Who Bought This Also Bought