Tapped Density Cylinder as per USP chapter 616, 100 mL, case of 2, 3.3 glass, Material: Glass, Size: 100mL, Base: Hexagon, Color: Clear, Graduation Color: Amber, Overall Dimension: 410mm L x 115mm W x 120mm H, icate Glass 3.3, Graduation Range: USP Type I, Class A
Catalog #: 76490-692
Supplier: Foxx Life Sciences
Customers Who Bought This Also Bought