Sentecdx Safety Glasses Clear Af Lens, Features rubberized temple tips and integrated nosepiece that fits comfortably with half mask respirators. Impact resistant polycarbonate lens with a scratch resistant coating provides 99.9% UV protection, Spring like hinges provide a perfect, comfortable fit
Catalog #: CA76492-856
Supplier: Dentec Safety Specialists
Customers Who Bought This Also Bought