Hartley Carb Safety Glasses Sm Mir Lens, Rubberized temple arms wrap gently around back of head behind ear providing a comfortable fit and ensures glasses stay in position, 6 base curved dual lens, vented temple arms, & fully adjustable soft silicone rubber nose pad with molded traction grips
Catalog #: CA76492-828
Supplier: Dentec Safety Specialists
Customers Who Bought This Also Bought